Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338026.1 | NP_001327857.1 | 3 - 4 | 444 | 63 | |
NM_001338027.1 | NP_001327858.1 | 2 - 3 | 710 | 63 | ||
NM_001338028.1 | NP_001327859.1 | 2 - 3 | 329 | 63 | ||
NM_001338029.1 | NP_001327860.1 | 2 - 3 | 473 | 63 | ||
NM_112143.4 | NP_187912.2 | 2 - 3 | 349 | 63 | ||
NM_180241.2 | NP_850572.1 | 2 - 3 | 329 | 63 | ||
GenBank mRNA | AK228498.1 | - | 2 - 3 | 129 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |