Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112355.3 | NP_566495.1 | 5 - 6 | 1293 | 63 | |
NM_180256.2 | NP_850587.1 | 5 - 6 | 1293 | 63 | ||
NM_202587.1 | NP_974316.1 | 5 - 6 | 1167 | 63 | ||
GenBank mRNA | AY136476.1 | - | 5 - 6 | 1077 | 63 | |
BT008826.1 | - | 3 - 4 | 848 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |