Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDA15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338339.1 | NP_001326233.1 | 2 - 3 | 306 | 108 | |
NM_001338340.1 | NP_001326232.1 | 2 - 3 | 306 | 108 | ||
NM_112737.3 | NP_566612.1 | 2 - 3 | 323 | 108 | ||
NM_180278.3 | NP_850609.2 | 2 - 3 | 306 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |