Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112862.4 | NP_188606.2 | 7 - 8 | 1369 | 99 | |
NM_180284.3 | NP_850615.2 | 7 - 8 | 1369 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |