Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035847.1 | NP_001030924.1 | 7 - 8 | 888 | 89 | |
NM_001197953.1 | NP_001184882.1 | 7 - 8 | 1290 | 89 | ||
NM_001331246.1 | NP_001320628.1 | 7 - 8 | 1290 | 89 | ||
NM_001331247.1 | NP_001320627.1 | 6 - 7 | 1171 | 89 | ||
NM_001331248.1 | NP_001320629.1 | 5 - 6 | 700 | 89 | ||
NM_099988.4 | NP_171614.1 | 7 - 8 | 971 | 89 | ||
NM_179237.1 | NP_849568.1 | 6 - 7 | 1081 | 89 | ||
GenBank mRNA | AK316728.1 | - | 6 - 7 | 1081 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |