Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_099996.4 | NP_171622.1 | 6 - 7 | 830 | 60 | |
NM_179239.1 | NP_849570.1 | 6 - 7 | 681 | 60 | ||
NM_179240.1 | NP_849571.1 | 6 - 7 | 681 | 60 | ||
GenBank mRNA | AF339147.1 | - | 6 - 7 | 568 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |