Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160826.1 | NP_001154298.1 | 5 - 6 | 858 | 64 | |
NM_099998.4 | NP_563619.1 | 4 - 5 | 774 | 64 | ||
GenBank mRNA | AF410307.1 | - | 4 - 5 | 694 | 64 | |
AY102133.1 | - | 4 - 5 | 530 | 64 | ||
AY102640.1 | - | 4 - 5 | 734 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |