Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP96A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342544.1 | NP_001328516.1 | 1 | 810 | 136 | |
NM_120110.1 | NP_195660.1 | 1 | 638 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |