Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120114.2 | NP_195664.2 | 1 - 2 | 422 | 172 | |
NM_202986.2 | NP_974715.1 | 2 - 3 | 288 | 172 | ||
GenBank mRNA | AK117791.1 | - | 1 - 2 | 422 | 172 | |
BT005291.1 | - | 1 - 2 | 195 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |