Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204040.1 | NP_001190969.1 | 5 - 6 | 509 | 82 | |
NM_120114.2 | NP_195664.2 | 4 - 5 | 611 | 82 | ||
NM_202986.2 | NP_974715.1 | 5 - 6 | 477 | 82 | ||
GenBank mRNA | AK117791.1 | - | 4 - 5 | 611 | 82 | |
BT005291.1 | - | 4 - 5 | 384 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |