Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204040.1 | NP_001190969.1 | 6 - 7 | 601 | 80 | |
NM_120114.2 | NP_195664.2 | 5 - 6 | 703 | 80 | ||
NM_202986.2 | NP_974715.1 | 6 - 7 | 569 | 80 | ||
GenBank mRNA | AK117791.1 | - | 5 - 6 | 703 | 80 | |
BT005291.1 | - | 5 - 6 | 476 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |