Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABH1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120152.4 | NP_195699.1 | 4 - 5 | 587 | 136 | |
GenBank mRNA | AF325109.1 | - | 4 - 5 | 394 | 136 | |
AF386952.1 | - | 4 - 5 | 537 | 136 | ||
BT000043.1 | - | 4 - 5 | 394 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |