Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203270.1 | NP_001190199.1 | 1 - 2 | 448 | 63 | |
NM_120205.5 | NP_195747.2 | 1 - 2 | 461 | 63 | ||
GenBank mRNA | AY557187.1 | - | 1 - 2 | 280 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |