Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120211.3 | NP_195753.1 | 2 - 3 | 770 | 91 | |
GenBank mRNA | AK227701.1 | - | 2 - 3 | 712 | 91 | |
BT006455.1 | - | 2 - 3 | 657 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |