Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342587.1 | NP_001330013.1 | 20 - 21 | 3578 | 66 | |
NM_001342588.1 | NP_001330014.1 | 21 - 22 | 3535 | 66 | ||
NM_120218.2 | NP_195760.1 | 21 - 22 | 3320 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |