Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342594.1 | NP_001331104.1 | 2 - 3 | 497 | 61 | |
NM_120227.4 | NP_568091.2 | 2 - 3 | 497 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |