Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120238.4 | NP_195780.1 | 6 - 7 | 1121 | 81 | |
GenBank mRNA | AF326869.1 | - | 6 - 7 | 733 | 81 | |
AF339691.1 | - | 6 - 7 | 664 | 81 | ||
AF412065.1 | - | 6 - 7 | 733 | 81 | ||
AK226433.1 | - | 6 - 7 | 736 | 81 | ||
X94248.1 | - | 702 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |