Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342645.1 | NP_001330945.1 | 7 - 8 | 906 | 71 | |
NM_120288.3 | NP_195830.1 | 8 - 9 | 1303 | 71 | ||
GenBank mRNA | AY050839.1 | - | 8 - 9 | 1064 | 71 | |
AY091166.1 | - | 8 - 9 | 945 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |