Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120291.4 | NP_195833.1 | 2 - 3 | 432 | 64 | |
GenBank mRNA | BT025628.1 | - | 2 - 3 | 322 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |