Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCI20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203276.1 | NP_001190205.1 | 6 - 7 | 831 | 82 | |
NM_120305.3 | NP_195847.1 | 5 - 6 | 915 | 82 | ||
GenBank mRNA | AF370554.1 | - | 5 - 6 | 695 | 82 | |
AY086132.1 | - | 5 - 6 | 723 | 82 | ||
BT008725.1 | - | 5 - 6 | 684 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |