Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342651.1 | NP_001318457.1 | 12 - 13 | 5425 | 84 | |
NM_001342652.1 | NP_001331954.1 | 12 - 13 | 5448 | 84 | ||
NM_120309.2 | NP_195851.2 | 11 - 12 | 5090 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |