Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342651.1 | NP_001318457.1 | 6 - 7 | 3090 | 74 | |
NM_001342652.1 | NP_001331954.1 | 6 - 7 | 3113 | 74 | ||
NM_120309.2 | NP_195851.2 | 5 - 6 | 2755 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |