Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120359.3 | NP_568107.1 | 3 - 4 | 782 | 63 | |
GenBank mRNA | AB046954.1 | - | 2 - 3 | 472 | 63 | |
AY039943.1 | - | 3 - 4 | 732 | 63 | ||
AY142560.1 | - | 2 - 3 | 472 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |