Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036746.2 | NP_001031823.1 | 6 - 7 | 923 | 94 | |
NM_120362.4 | NP_568108.1 | 6 - 7 | 923 | 94 | ||
NM_180425.2 | NP_850756.1 | 6 - 7 | 865 | 94 | ||
GenBank mRNA | AJ937209.1 | - | 6 - 7 | 777 | 94 | |
AK317012.1 | - | 6 - 7 | 865 | 94 | ||
AY519514.1 | - | 6 - 7 | 777 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |