Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120393.6 | NP_195935.2 | 2 - 3 | 1015 | 87 | |
GenBank mRNA | AJ621494.1 | - | 2 - 3 | 587 | 87 | |
AJ630493.1 | - | 2 - 3 | 587 | 87 | ||
AY568665.1 | - | 2 - 3 | 587 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |