Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P58IPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120394.4 | NP_195936.1 | 2 - 3 | 485 | 140 | |
GenBank mRNA | AY085329.1 | - | 2 - 3 | 438 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |