Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P58IPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120394.4 | NP_195936.1 | 8 - 9 | 1442 | 66 | |
GenBank mRNA | AY085329.1 | - | 8 - 9 | 1395 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |