Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036747.1 | NP_001031824.1 | 2 | 1501 | 137 | |
NM_120402.3 | NP_568112.2 | 1 | 1483 | 137 | ||
NM_180698.3 | NP_851029.1 | 2 | 1326 | 137 | ||
GenBank mRNA | AK317101.1 | - | 2 | 1152 | 137 | |
BT002420.1 | - | 2 | 1168 | 137 |
Gene Symbol: AT1G54410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK230245.1 | - | 80 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |