Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120404.3 | NP_195946.2 | 2 - 3 | 332 | 109 | |
GenBank mRNA | AY065128.1 | - | 2 - 3 | 332 | 109 | |
AY081592.1 | - | 2 - 3 | 246 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |