Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCSLA09 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120457.4 | NP_195996.1 | 8 - 9 | 1983 | 111 | |
GenBank mRNA | AY060563.1 | - | 8 - 9 | 1903 | 111 | |
AY149935.1 | - | 8 - 9 | 1440 | 111 | ||
KJ138709.1 | - | 8 - 9 | 1440 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |