Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSRP54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120476.5 | NP_196014.1 | 7 - 8 | 976 | 120 | |
GenBank mRNA | AY050999.1 | - | 7 - 8 | 872 | 120 | |
BT000976.1 | - | 7 - 8 | 808 | 120 | ||
Z21970.1 | - | 831 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |