Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127444.4 | NP_179478.4 | 7 - 8 | 1252 | 72 | |
GenBank mRNA | AY924744.1 | - | 4 - 5 | 941 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |