Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127476.2 | NP_179509.1 | 8 - 9 | 1698 | 70 | |
GenBank mRNA | AF486619.1 | - | 8 - 9 | 1698 | 70 | |
FJ708695.1 | - | 8 - 9 | 1647 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |