Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2-BETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202625.1 | NP_001189554.1 | 3 - 4 | 313 | 71 | |
NM_127527.4 | NP_179559.2 | 3 - 4 | 356 | 71 | ||
GenBank mRNA | AF541915.2 | - | 3 - 4 | 266 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |