Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335672.1 | NP_001325368.1 | 7 - 8 | 1019 | 97 | |
NM_001335673.1 | NP_001325369.1 | 7 - 8 | 1019 | 97 | ||
NM_001335674.1 | NP_001325367.1 | 7 - 8 | 1026 | 97 | ||
NM_127588.3 | NP_179619.2 | 7 - 8 | 828 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |