Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127618.4 | NP_565477.1 | 15 - 16 | 1975 | 74 | |
GenBank mRNA | AF360216.1 | - | 15 - 16 | 1927 | 74 | |
AY040061.1 | - | 15 - 16 | 1726 | 74 | ||
AY230828.1 | - | 15 - 16 | 1810 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |