Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127618.4 | NP_565477.1 | 7 - 8 | 940 | 138 | |
GenBank mRNA | AF360216.1 | - | 7 - 8 | 892 | 138 | |
AY040061.1 | - | 7 - 8 | 691 | 138 | ||
AY230828.1 | - | 7 - 8 | 775 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |