Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127645.4 | NP_179673.1 | 1 - 2 | 225 | 111 | |
GenBank mRNA | DQ446535.1 | - | 1 - 2 | 93 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |