Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202636.1 | NP_001189565.1 | 8 - 9 | 1034 | 78 | |
NM_001202637.1 | NP_001189566.1 | 9 - 10 | 1148 | 78 | ||
NM_127668.4 | NP_565495.1 | 8 - 9 | 1116 | 78 | ||
GenBank mRNA | AB102951.1 | - | 8 - 9 | 759 | 78 | |
AJ617630.1 | - | 8 - 9 | 1012 | 78 | ||
AY045836.1 | - | 8 - 9 | 1019 | 78 | ||
AY087925.1 | - | 8 - 9 | 1014 | 78 | ||
BT004371.1 | - | 8 - 9 | 759 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |