Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036305.1 | NP_001031382.1 | 6 - 7 | 1355 | 74 | |
NM_001202638.2 | NP_001189567.1 | 6 - 7 | 1410 | 74 | ||
NM_127677.3 | NP_179703.2 | 6 - 7 | 1399 | 74 | ||
GenBank mRNA | BT015875.1 | - | 6 - 7 | 1379 | 74 | |
BT020210.1 | - | 6 - 7 | 1322 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |