Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084462.1 | NP_001077931.1 | 3 - 4 | 517 | 95 | |
NM_001335732.1 | NP_001323967.1 | 5 - 6 | 679 | 95 | ||
NM_127687.4 | NP_179713.1 | 4 - 5 | 640 | 95 | ||
GenBank mRNA | AF247559.1 | - | 4 - 5 | 477 | 95 | |
AF378898.1 | - | 4 - 5 | 466 | 95 | ||
AY052748.1 | - | 4 - 5 | 417 | 95 | ||
AY087893.1 | - | 4 - 5 | 511 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |