Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339193.1 | NP_001326667.1 | 7 - 8 | 1407 | 121 | |
NM_114379.3 | NP_566874.1 | 7 - 8 | 1261 | 121 | ||
NM_180327.2 | NP_850658.1 | 7 - 8 | 1261 | 121 | ||
GenBank mRNA | AY039602.1 | - | 7 - 8 | 1258 | 121 | |
AY081726.1 | - | 6 - 7 | 1161 | 121 | ||
KJ138805.1 | - | 6 - 7 | 1161 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |