Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114613.3 | NP_190329.2 | 10 - 11 | 1336 | 87 | |
NM_180335.1 | NP_850666.1 | 10 - 11 | 1336 | 87 | ||
GenBank mRNA | AK226195.1 | - | 10 - 11 | 1341 | 87 | |
BT015353.1 | - | 10 - 11 | 1241 | 87 | ||
BT015887.1 | - | 10 - 11 | 1238 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |