Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114613.3 | NP_190329.2 | 3 - 4 | 772 | 102 | |
NM_180335.1 | NP_850666.1 | 3 - 4 | 772 | 102 | ||
GenBank mRNA | AK226195.1 | - | 3 - 4 | 777 | 102 | |
BT015353.1 | - | 3 - 4 | 677 | 102 | ||
BT015887.1 | - | 3 - 4 | 674 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |