Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114680.4 | NP_190394.3 | 5 - 6 | 860 | 84 | |
GenBank mRNA | AY136413.1 | - | 5 - 6 | 776 | 84 | |
BT012191.1 | - | 5 - 6 | 719 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |