Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115261.4 | NP_566993.1 | 13 - 14 | 1377 | 91 | |
NM_180370.3 | NP_850701.1 | 12 - 13 | 1407 | 91 | ||
GenBank mRNA | AY056265.1 | - | 13 - 14 | 1309 | 91 | |
AY096635.1 | - | 13 - 14 | 1156 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |