Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035821.1 | NP_001030898.1 | 7 - 8 | 727 | 131 | |
NM_001203210.1 | NP_001190139.1 | 7 - 8 | 963 | 131 | ||
NM_115862.3 | NP_567089.1 | 7 - 8 | 754 | 131 | ||
NM_180394.2 | NP_850725.1 | 6 - 7 | 894 | 131 | ||
GenBank mRNA | AF300880.1 | - | 6 - 7 | 819 | 131 | |
AY084710.1 | - | 7 - 8 | 753 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |