Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116025.4 | NP_567115.1 | 5 - 6 | 1143 | 70 | |
NM_180402.2 | NP_850733.1 | 5 - 6 | 1143 | 70 | ||
GenBank mRNA | AF292397.1 | - | 5 - 6 | 937 | 70 | |
BT030347.1 | - | 5 - 6 | 861 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |