Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180415.3 | NP_850746.1 | 3 - 4 | 615 | 67 | |
GenBank mRNA | AF347077.1 | - | 3 - 4 | 309 | 67 | |
AK228306.1 | - | 3 - 4 | 502 | 67 | ||
BT024657.1 | - | 3 - 4 | 217 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |