Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203285.1 | NP_001190214.1 | 6 - 7 | 1024 | 95 | |
NM_180426.3 | NP_850757.1 | 6 - 7 | 843 | 95 | ||
GenBank mRNA | AB493733.1 | - | 5 - 6 | 583 | 95 | |
AJ294532.1 | - | 5 - 6 | 583 | 95 | ||
AK228148.1 | - | 6 - 7 | 824 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |